Learn More
Grainger InsulMultitapCon PVC 6 AWG 350 kcmil
Catalog No. 502750629
Encompass_Preferred
The Grainger Approved InsulMultitapCon PVC 6 AWG 350 kcmil is a PVC insulated multitap connector for 6 AWG 350 kcmil copper wire. It is rated for 600 volts and has a temperature rating of 105°C. The connector has 4 ports and is made of high quality thermoplastic with UL 94 V-0 flammability rating. It meets UL 486 standards and is RoHS compliant. This multitap connector provides a reliable electrical connection for branching and extending 6 AWG 350 kcmil building wire.
Catalog No. 50-275-0629 Supplier Grainger Supplier No. 14V964
The Grainger Approved InsulMultitapCon PVC 6 AWG 350 kcmil is a PVC insulated multitap connector for 6 AWG 350 kcmil copper wire. It is rated for 600 volts and has a temperature rating of 105°C. The connector has 4 ports and is made of high quality thermoplastic with UL 94 V-0 flammability rating. It meets UL 486 standards and is RoHS compliant. This multitap connector provides a reliable electrical connection for branching and extending 6 AWG 350 kcmil building wire.