Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

InSure™ FIT™ Immunochemical Test Kit

Catalog No. 22823006
Change view
Click to view available options
Product Type:
Developer Kit
FOBT Control
InSure FIT Combo Kit, 25 Pack and 10 Pack Patient Collection Kit
InSure FIT Combo Kit, 25 Patient Collection Kit
InSure FIT Patient Collection Kit
QC Pack
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-823-006 FOBT Control
22-823-001 Developer Kit
22-823-003 InSure FIT Patient Collection Kit
22-823-004 InSure FIT Combo Kit, 25 Patient Collection Kit
22-823-005 InSure FIT Combo Kit, 25 Pack and 10 Pack Patient Collection Kit
22-823-007 QC Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-823-006 Supplier InSure™ Supplier No. 16800
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The InSure FIT test and its unique Blue Brush Method make it easier to screen for colorectal cancer

Clinical studies have shown that the InSure FIT test is highly sensitive for detecting blood in and around the stool, a key indicator of abnormalities in the colon and rectum that doctors use to help screen for colorectal cancer.

InSure FIT Test Kit

  • Easy to Use - patients only need to gently brush the surface of the stool in water for about 5 seconds with the convenient, long-handled blue brush
  • Convenient - samples are collected in the privacy of patient's home, then submitted to the doctor's office or lab for analysis
  • Specific - does not react with non-human vitamins, drugs or peroxidase from food sources, as Insure FIT is specific for the globin portion of human hemoglobin

  • CPT Code 82274 and Medicare Code G0328QW
  • No fecal handling – Blue Brush method
  • No dietary or medicinal restrictions
  • 87% sensitivity for colorectal cancer1, 2
  • Study demonstrated increased patient compliance1
  • Room temperature storage
  • Two samples on one test card
  • Internal controls
  • CLIA waived

Specifications

Includes 2 bottle each, positive and negative, 1.5mL per bottle
Quantity 1 Kit
Sample Volume 1.5 mL
Form Liquid
For Use With (Application) Detection of Blood in and around the stool, helps screen for colorectal cancer
Product Type FOBT Control
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.