Learn More
InSure™ FIT™ Immunochemical Test Kit
Description
Clinical studies have shown that the InSure FIT test is highly sensitive for detecting blood in and around the stool, a key indicator of abnormalities in the colon and rectum that doctors use to help screen for colorectal cancer.
InSure FIT Test Kit
- Easy to Use - patients only need to gently brush the surface of the stool in water for about 5 seconds with the convenient, long-handled blue brush
- Convenient - samples are collected in the privacy of patient's home, then submitted to the doctor's office or lab for analysis
- Specific - does not react with non-human vitamins, drugs or peroxidase from food sources, as Insure FIT is specific for the globin portion of human hemoglobin
- CPT Code 82274 and Medicare Code G0328QW
- No fecal handling – Blue Brush method
- No dietary or medicinal restrictions
- 87% sensitivity for colorectal cancer1, 2
- Study demonstrated increased patient compliance1
- Room temperature storage
- Two samples on one test card
- Internal controls
- CLIA waived
Specifications
Specifications
| Cross Reactivity | Human |
| Includes | 25 Test strip, 9mL bottle of reagent |
| Quantity | 1 Kit |
| Form | Liquid |
| For Use With (Application) | Detection of Blood in and around the stool, helps screen for colorectal cancer |
| Product Type | Developer Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.