Learn More
Hanna Pharmaceutical Supply Co., Inc. BD INSYTE™ AUTOGUARD™ IV Catheter, 20G x 1", Pink, BC Shielded, 50/bx, 4 bx/cs
Catalog No. NC2144496
Encompass_Preferred
Shielded IV Catheter with Blood Control Technology, BD Vialon™ catheter biomaterial and BD Instaflash™ needle technology. IV Catheter, 20G x 1", Pink, BC Shielded, 50/bx, 4 bx/cs (Continental US Only)
This item is not returnable. View return policy
Catalog No. NC2144496 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. BD382533
Shielded IV Catheter with Blood Control Technology, BD Vialon™ catheter biomaterial and BD Instaflash™ needle technology. IV Catheter, 20G x 1", Pink, BC Shielded, 50/bx, 4 bx/cs (Continental US Only)