Learn More
Medchemexpress LLC HY-12222 500mg , INT-747 Obeticholic acid;6-ECDCA;6-Ethylchenodeoxycholic acid CAS:459789-99-2 Purity:98%
SDP
Catalog No. 501492320
Encompass_Preferred
Medchemexpress, HY-12222 500mg INT-747 Obeticholic acid;6-ECDCA;6-Ethylchenodeoxycholic acid CAS:459789-99-2 Formula:C26H44O4 EC50: 99 nM (FXR) Purity:98% INT-747 is a potent and selective FXR agonist ( EC 50 =99 nM) endowed with anticholestatic activity. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2320 Supplier Medchemexpress LLC Supplier No. HY12222500MG
Medchemexpress, HY-12222 500mg INT-747 Obeticholic acid;6-ECDCA;6-Ethylchenodeoxycholic acid CAS:459789-99-2 Formula:C26H44O4 EC50: 99 nM (FXR) Purity:98% INT-747 is a potent and selective FXR agonist ( EC 50 =99 nM) endowed with anticholestatic activity. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.