Learn More
Medchemexpress LLC Int-767 | 1000403-03-1 | 99.8% | 494.66 g/mol | C25H43NaO6S | 10MG
SDP
Catalog No. 5000238974
Encompass_Preferred
INT-767 is a semisynthetic dual farnesoid X receptor (FXR) and TGR5 (GPBAR1) agonist used in preclinical research to modulate bile acid signaling, liver, and metabolic pathways. The compound shows mean EC50s of 30 nM for FXR and 630 nM for TGR5, and is supplied as the sodium salt with high reported purity.
Catalog No. 50-002-38974 Supplier Medchemexpress LLC Supplier No. HY1243410MG
INT-767 is a semisynthetic dual farnesoid X receptor (FXR) and TGR5 (GPBAR1) agonist used in preclinical research to modulate bile acid signaling, liver, and metabolic pathways. The compound shows mean EC50s of 30 nM for FXR and 630 nM for TGR5, and is supplied as the sodium salt with high reported purity.