Learn More
Selleck Chemical LLC INT-777
SDP
Catalog No. 5000000868
Encompass_Preferred
INT-777 (S-EMCA) is a potent and selective agonist of TGR5(GPCR19) with EC50 of 0 18 M 3 9 M and 1 45 M for TGR5 TGR5(N76A) and TGR5(Y89F) respectively
Catalog No. 50-000-00868 Supplier Selleck Chemical LLC Supplier No. S82235MG
INT-777 (S-EMCA) is a potent and selective agonist of TGR5(GPCR19) with EC50 of 0 18 M 3 9 M and 1 45 M for TGR5 TGR5(N76A) and TGR5(Y89F) respectively