Learn More
Medchemexpress LLC HY-15677A 2mg Medchemexpress, INT-777 (R-enantiomer) CAS:1198786-98-9 Purity:>98%
SDP
Catalog No. 501932753
Encompass_Preferred
Medchemexpress, HY-15677A 2mg INT-777 (R-enantiomer) CAS:1198786-98-9 INT-777 (R-enantiomer) is the R-enantiomer of INT-777, with EC 50 of 4.79 μM for TGR5, and less potent than INT-777. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2753 Supplier Medchemexpress LLC Supplier No. HY15677A2MG
Medchemexpress, HY-15677A 2mg INT-777 (R-enantiomer) CAS:1198786-98-9 INT-777 (R-enantiomer) is the R-enantiomer of INT-777, with EC 50 of 4.79 μM for TGR5, and less potent than INT-777. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.