Learn More
Abcam INT dye, tetrazolium dye precursor, 250MG
Catalog No. 502704704
Encompass_Preferred
MW 505.71 g/mol, Purity >98%. Tetrazolium dye precursor that forms a purple formazan dye on reduction. Used as an electron acceptor for the colorimetric assay of dehydrogenase activity. Also used for western blot staining and as a staining agent for soft agar colony formation assays.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4704 Supplier Abcam Supplier No. AB146338250MG
MW 505.71 g/mol, Purity >98%. Tetrazolium dye precursor that forms a purple formazan dye on reduction. Used as an electron acceptor for the colorimetric assay of dehydrogenase activity. Also used for western blot staining and as a staining agent for soft agar colony formation assays.
The product is subject to the following: Abcam Restricted Use Statement