Learn More
Abcam INT dye, tetrazolium dye precursor, 5G
Catalog No. 502704703
Encompass_Preferred
MW 505.71 g/mol, Purity >98%. Tetrazolium dye precursor that forms a purple formazan dye on reduction. Used as an electron acceptor for the colorimetric assay of dehydrogenase activity. Also used for western blot staining and as a staining agent for soft agar colony formation assays.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4703 Supplier Abcam Supplier No. AB1463385G
MW 505.71 g/mol, Purity >98%. Tetrazolium dye precursor that forms a purple formazan dye on reduction. Used as an electron acceptor for the colorimetric assay of dehydrogenase activity. Also used for western blot staining and as a staining agent for soft agar colony formation assays.
The product is subject to the following: Abcam Restricted Use Statement