Learn More
INTACT GENOMICS INC IG Autoinduction Medium, 1L
Catalog No. NC3998435
Encompass
This autoinduction medium is designed to simplify and enhance the production of T7 promoter-regulated recombinant proteins in E. coli. It eliminates the need for manual induction and frequent optical density monitoring, providing a “set-and-forget” approach. This results in repeatable, high-yield protein expression and high cell densities.
  • Eliminates frequent OD600 monitoring or manual inducer addition
  • Provides reliable and consistent protein expression
  • Reduces issues like leaky expression, vector instability, and challenges with toxic proteins
  • Promotes high cell density and robust protein expression
  • Compatible with other DE3 strains and media
  • Automates protein expression after inoculation
Catalog No. NC3998435 Supplier INTACT GENOMICS INC Supplier No. 17221L
This autoinduction medium is designed to simplify and enhance the production of T7 promoter-regulated recombinant proteins in E. coli. It eliminates the need for manual induction and frequent optical density monitoring, providing a “set-and-forget” approach. This results in repeatable, high-yield protein expression and high cell densities.
  • Eliminates frequent OD600 monitoring or manual inducer addition
  • Provides reliable and consistent protein expression
  • Reduces issues like leaky expression, vector instability, and challenges with toxic proteins
  • Promotes high cell density and robust protein expression
  • Compatible with other DE3 strains and media
  • Automates protein expression after inoculation