Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INTACT GENOMICS INC IG Autoinduction Medium, 40g+50mL

Catalog No. NC3998437
Encompass

Intact Genomics Autoinduction Medium facilitates high-yield growth of E. coli and robust expression of T7 promoter-regulated recombinant proteins. This medium eliminates the need for labor-intensive steps such as frequent optical density (OD) monitoring or the manual addition of induction components like IPTG. It supports automatic protein expression upon inoculation with a single colony or starter culture.

  • Simple and fast: Eliminates the need for frequent OD600 monitoring or manual inducer addition, saving time.
  • Repeatable: Provides reliable and consistent protein expression by mitigating leaky expression, vector instability, and issues with toxic proteins.
  • High-yield: Facilitates high cell density and robust protein expression.
  • Flexibility: Compatible with other DE3 strains and media, allowing for seamless integration into existing workflows.

Catalog No. NC3998437 Supplier INTACT GENOMICS INC Supplier No. 1722PD1L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Intact Genomics Autoinduction Medium facilitates high-yield growth of E. coli and robust expression of T7 promoter-regulated recombinant proteins. This medium eliminates the need for labor-intensive steps such as frequent optical density (OD) monitoring or the manual addition of induction components like IPTG. It supports automatic protein expression upon inoculation with a single colony or starter culture.

  • Simple and fast: Eliminates the need for frequent OD600 monitoring or manual inducer addition, saving time.
  • Repeatable: Provides reliable and consistent protein expression by mitigating leaky expression, vector instability, and issues with toxic proteins.
  • High-yield: Facilitates high cell density and robust protein expression.
  • Flexibility: Compatible with other DE3 strains and media, allowing for seamless integration into existing workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.