Learn More
INTACT GENOMICS INC IG C41(DE3) Chemically Competent Cells 24x50µl
Catalog No. NC3998272
Encompass
IG C41(DE3) Chemically Competent Cells are engineered for transformation and routine recombinant protein expression from IPTG-inducible T7 promoter vectors. This C41(DE3) strain improves tolerance to toxic protein expression and reduces cell death, ensuring reliable T7-driven performance.
  • Engineered for improved tolerance to toxic protein expression and reduced cell death.
  • Facilitates routine recombinant protein expression from IPTG-inducible T7 promoter vectors.
  • Enables efficient expression of toxic proteins from diverse organisms.
  • T7 expression strain.
  • Suitable for expression of toxic genes.
  • Chemically competent cell type, E. coli species, C41(DE3) strain.
  • Available in tube format.
  • Transformation efficiency: ≥ 3.0 × 107 cfu/μg pUC19 DNA.
  • Blue/white screening is not supported.
  • Shipped on dry ice.
Catalog No. NC3998272 Supplier INTACT GENOMICS INC Supplier No. 104324
IG C41(DE3) Chemically Competent Cells are engineered for transformation and routine recombinant protein expression from IPTG-inducible T7 promoter vectors. This C41(DE3) strain improves tolerance to toxic protein expression and reduces cell death, ensuring reliable T7-driven performance.
  • Engineered for improved tolerance to toxic protein expression and reduced cell death.
  • Facilitates routine recombinant protein expression from IPTG-inducible T7 promoter vectors.
  • Enables efficient expression of toxic proteins from diverse organisms.
  • T7 expression strain.
  • Suitable for expression of toxic genes.
  • Chemically competent cell type, E. coli species, C41(DE3) strain.
  • Available in tube format.
  • Transformation efficiency: ≥ 3.0 × 107 cfu/μg pUC19 DNA.
  • Blue/white screening is not supported.
  • Shipped on dry ice.