Learn More
INTACT GENOMICS INC ig ccdB Resist Chemically Competent Cells, 24x50μl
Catalog No. NC3998348
Encompass
These ccdB Resist cells are capable of propagating plasmids containing the ccdB gene. Chemically competent E. coli cells offer high efficiency transformation for various applications, including cloning and sub-cloning, leading to increased experimental success.
  • Resistance to the ccdB gene product, as well as the T1 and T5 phage.
  • High transformation efficiency.
  • EndA1 mutation increases plasmid yield significantly.
  • Capable of blue/white screening.
Catalog No. NC3998348 Supplier INTACT GENOMICS INC Supplier No. 106924
These ccdB Resist cells are capable of propagating plasmids containing the ccdB gene. Chemically competent E. coli cells offer high efficiency transformation for various applications, including cloning and sub-cloning, leading to increased experimental success.
  • Resistance to the ccdB gene product, as well as the T1 and T5 phage.
  • High transformation efficiency.
  • EndA1 mutation increases plasmid yield significantly.
  • Capable of blue/white screening.