Learn More
INTACT GENOMICS INC IG C41(DE3)pLysS Electro Competent Cells 24x50µl
Catalog No. NC3998378
Encompass
Intact Genomics 5 alpha chemically competent E. coli cells are designed for high-efficiency transformation in various applications, including cloning and subcloning. These cells offer at least twice the transformation efficiency of their nearest competitor. They provide reliable, high-efficiency transformation for routine cloning applications, consistent performance, and fast recovery, which helps to streamline everyday workflows.
  • Suitable for high efficiency transformation in a wide variety of applications such as cloning and sub-cloning.
  • At least twice the transformation efficiency of the nearest competitor.
  • Deliver consistent performance and fast recovery.
  • Helps streamline everyday workflows.
Catalog No. NC3998378 Supplier INTACT GENOMICS INC Supplier No. 124548
Intact Genomics 5 alpha chemically competent E. coli cells are designed for high-efficiency transformation in various applications, including cloning and subcloning. These cells offer at least twice the transformation efficiency of their nearest competitor. They provide reliable, high-efficiency transformation for routine cloning applications, consistent performance, and fast recovery, which helps to streamline everyday workflows.
  • Suitable for high efficiency transformation in a wide variety of applications such as cloning and sub-cloning.
  • At least twice the transformation efficiency of the nearest competitor.
  • Deliver consistent performance and fast recovery.
  • Helps streamline everyday workflows.