Learn More
INTACT GENOMICS INC IG Autoinduction DE3 Chemically Competent Cells, 1x96 well plate (20µl/well)
Catalog No. NC3998330
Encompass
Autoinduction DE3 Chemically Competent cells are designed for transformation and routine recombinant protein expression from a vector under an IPTG-inducible T7 promoter. This DE3 lysogen strain features a chromosomally integrated IPTG-inducible promoter, enabling T7 RNA polymerase expression via IPTG induction and autoinduction. This approach simplifies autoinduction, potentially boosting yields of cells and protein for various targets. This specific product is provided in a 1x96 well plate format.
  • T7 expression strain.
  • Optimized for autoinduction.
  • Suitable for expression of toxic genes.
  • Tetracycline resistant (12.5 μg/mL).
  • DE3 lysogen with chromosomally integrated IPTG-inducible promoter.
  • Simple, less hands-on autoinduction process.
  • Transformation efficiency: ≥ 3.0 x 107 cfu/μg pUC19 DNA.
  • Packaged in a 1x96 well plate, 20 μL/well.
Catalog No. NC3998330 Supplier INTACT GENOMICS INC Supplier No. 106596
Autoinduction DE3 Chemically Competent cells are designed for transformation and routine recombinant protein expression from a vector under an IPTG-inducible T7 promoter. This DE3 lysogen strain features a chromosomally integrated IPTG-inducible promoter, enabling T7 RNA polymerase expression via IPTG induction and autoinduction. This approach simplifies autoinduction, potentially boosting yields of cells and protein for various targets. This specific product is provided in a 1x96 well plate format.
  • T7 expression strain.
  • Optimized for autoinduction.
  • Suitable for expression of toxic genes.
  • Tetracycline resistant (12.5 μg/mL).
  • DE3 lysogen with chromosomally integrated IPTG-inducible promoter.
  • Simple, less hands-on autoinduction process.
  • Transformation efficiency: ≥ 3.0 x 107 cfu/μg pUC19 DNA.
  • Packaged in a 1x96 well plate, 20 μL/well.