Learn More
INTACT GENOMICS INC IG C41(DE3)pLysS Chemically Competent Cells 24x50µl
Catalog No. NC3998287
Encompass
These C41(DE3)pLysS Chemically Competent Cells are engineered for routine recombinant protein expression driven by an IPTG-inducible T7 promoter. Derived from BL21(DE3), this strain is designed to prevent cell death associated with toxic protein expression, making it highly effective for expressing toxic proteins from various organisms. The C41(DE3)pLysS strain also includes a chloramphenicol-resistant plasmid encoding T7 lysozyme, which suppresses basal T7 RNA polymerase expression before induction, stabilizing recombinants of toxic proteins. Its efficacy has been validated in over 400 publications.
  • Designed for IPTG-inducible T7 systems
  • Improves tolerance to toxic proteins
  • pLysS plasmid suppresses basal expression for greater stability
  • Chloramphenicol-resistant (12.5 μg/mL)
  • T7 expression strain
Catalog No. NC3998287 Supplier INTACT GENOMICS INC Supplier No. 104524
These C41(DE3)pLysS Chemically Competent Cells are engineered for routine recombinant protein expression driven by an IPTG-inducible T7 promoter. Derived from BL21(DE3), this strain is designed to prevent cell death associated with toxic protein expression, making it highly effective for expressing toxic proteins from various organisms. The C41(DE3)pLysS strain also includes a chloramphenicol-resistant plasmid encoding T7 lysozyme, which suppresses basal T7 RNA polymerase expression before induction, stabilizing recombinants of toxic proteins. Its efficacy has been validated in over 400 publications.
  • Designed for IPTG-inducible T7 systems
  • Improves tolerance to toxic proteins
  • pLysS plasmid suppresses basal expression for greater stability
  • Chloramphenicol-resistant (12.5 μg/mL)
  • T7 expression strain