Learn More
INTACT GENOMICS INC IG C43(DE3)pLysS Chemically Competent Cells 24x50µl
Catalog No. NC3998309
Encompass
IG™ C43(DE3)pLysS Chemically Competent Cells are engineered for high-efficiency transformation and stable expression of toxic proteins in IPTG-inducible T7 systems. Derived from BL21(DE3), these cells offer improved tolerance to toxic proteins. A pLysS plasmid helps stabilize challenging constructs by suppressing basal expression and provides chloramphenicol resistance.
  • Engineered for high-efficiency transformation & stable expression
  • Derived from BL21(DE3) for improved tolerance
  • pLysS plasmid for basal expression suppression
  • Effective for diverse organisms
  • Chloramphenicol-resistant
  • Validated in >400 publications
  • Genetic mutations identified
  • T7 expression strain
Catalog No. NC3998309 Supplier INTACT GENOMICS INC Supplier No. 104924
IG™ C43(DE3)pLysS Chemically Competent Cells are engineered for high-efficiency transformation and stable expression of toxic proteins in IPTG-inducible T7 systems. Derived from BL21(DE3), these cells offer improved tolerance to toxic proteins. A pLysS plasmid helps stabilize challenging constructs by suppressing basal expression and provides chloramphenicol resistance.
  • Engineered for high-efficiency transformation & stable expression
  • Derived from BL21(DE3) for improved tolerance
  • pLysS plasmid for basal expression suppression
  • Effective for diverse organisms
  • Chloramphenicol-resistant
  • Validated in >400 publications
  • Genetic mutations identified
  • T7 expression strain