Learn More
INTACT GENOMICS INC IG C43(DE3)pLysS Chemically Competent Cells 6x50µl
Catalog No. NC3998304
Encompass
These chemically competent cells are engineered for high-efficiency transformation and stable expression of toxic or difficult proteins within IPTG-inducible T7 systems. Derived from BL21(DE3), this strain offers improved tolerance to toxic proteins, while the pLysS plasmid works to suppress basal expression, thereby stabilizing challenging constructs.
  • T7 expression strain
  • Selected for expression of toxic proteins
  • Suitable for expression of toxic genes
  • Chloramphenicol-resistant (12.5 μg/ml)
  • High-efficiency transformation
  • Improved tolerance to toxic proteins
  • Suppresses basal expression of T7 RNA polymerase
Catalog No. NC3998304 Supplier INTACT GENOMICS INC Supplier No. 104906
These chemically competent cells are engineered for high-efficiency transformation and stable expression of toxic or difficult proteins within IPTG-inducible T7 systems. Derived from BL21(DE3), this strain offers improved tolerance to toxic proteins, while the pLysS plasmid works to suppress basal expression, thereby stabilizing challenging constructs.
  • T7 expression strain
  • Selected for expression of toxic proteins
  • Suitable for expression of toxic genes
  • Chloramphenicol-resistant (12.5 μg/ml)
  • High-efficiency transformation
  • Improved tolerance to toxic proteins
  • Suppresses basal expression of T7 RNA polymerase