Learn More
INTACT GENOMICS INC IG C43(DE3) Chemically Competent Cells 24x50µl
Catalog No. NC3998300
Encompass
These C43(DE3) Chemically Competent Cells are optimized for the expression of toxic and difficult proteins using an IPTG-inducible T7 promoter. Derived from BL21(DE3) and selected from C41(DE3) survivor cells, C43(DE3) carries mutations to prevent cell death during recombinant toxic protein expression. This strain is effective for expressing toxic proteins from a wide range of organisms, validated by patent and publications.
  • T7 expression strain
  • Selected for expression of toxic proteins
  • Suitable for expression of toxic genes
  • Chemically competent cell type
  • E. coli species
  • C43(DE3) strain
  • Tubes format
  • Transformation efficiency ≥ 3.0 × 107 cfu/µg pUC19 DNA
  • No blue/white screening
  • Dry ice shipping condition
Catalog No. NC3998300 Supplier INTACT GENOMICS INC Supplier No. 104724
These C43(DE3) Chemically Competent Cells are optimized for the expression of toxic and difficult proteins using an IPTG-inducible T7 promoter. Derived from BL21(DE3) and selected from C41(DE3) survivor cells, C43(DE3) carries mutations to prevent cell death during recombinant toxic protein expression. This strain is effective for expressing toxic proteins from a wide range of organisms, validated by patent and publications.
  • T7 expression strain
  • Selected for expression of toxic proteins
  • Suitable for expression of toxic genes
  • Chemically competent cell type
  • E. coli species
  • C43(DE3) strain
  • Tubes format
  • Transformation efficiency ≥ 3.0 × 107 cfu/µg pUC19 DNA
  • No blue/white screening
  • Dry ice shipping condition