Learn More
INTACT GENOMICS INC DL39(DE3) Chemically Competent Cells 6x200µl
Catalog No. NC3998451
Encompass
Intact Genomics ccdB Resist Chemically Competent E. coli cells are designed for high efficiency transformation in various applications, including cloning and sub-cloning. These cells are capable of propagating plasmids that contain the ccdB gene, offering excellent transformation efficiency to enhance experimental success.
  • Resistance to the ccdB gene product, as well as the T1 and T5 phage.
  • High transformation efficiency.
  • EndA1 mutation increases plasmid yield significantly.
  • Capable of blue/white screening.
Catalog No. NC3998451 Supplier INTACT GENOMICS INC Supplier No. 106424
Intact Genomics ccdB Resist Chemically Competent E. coli cells are designed for high efficiency transformation in various applications, including cloning and sub-cloning. These cells are capable of propagating plasmids that contain the ccdB gene, offering excellent transformation efficiency to enhance experimental success.
  • Resistance to the ccdB gene product, as well as the T1 and T5 phage.
  • High transformation efficiency.
  • EndA1 mutation increases plasmid yield significantly.
  • Capable of blue/white screening.