Learn More
INTACT GENOMICS INC Recombineering Chemically Competent Cells 6x50l
Catalog No. NC3998335
Encompass
Intact Genomics (IG™) Recombineering Chemically Competent Cells enable high-efficiency transformation for DNA, plasmid, and BAC engineering, and homologous recombination. Derived from SW102 (a 10B background strain), they feature a defective lambda prophage and a galK deletion for efficient DNA/BAC modification via galK positive/negative selection. The cells are tetracycline resistant (5 μg/mL).
  • High transformation efficiency.
  • Supports DNA, plasmid, and BAC engineering, plus homologous recombination.
  • Enables precise modification using galK positive/negative selection.
  • Derived from SW102 (10B background).
  • Heat-inducible prophage with red genes (exo, bet, gam), repressed at 32 °C and induced at 42 °C.
  • Tetracycline resistant (TetR: 5 μg/ml).
Catalog No. NC3998335 Supplier INTACT GENOMICS INC Supplier No. 106606
Intact Genomics (IG™) Recombineering Chemically Competent Cells enable high-efficiency transformation for DNA, plasmid, and BAC engineering, and homologous recombination. Derived from SW102 (a 10B background strain), they feature a defective lambda prophage and a galK deletion for efficient DNA/BAC modification via galK positive/negative selection. The cells are tetracycline resistant (5 μg/mL).
  • High transformation efficiency.
  • Supports DNA, plasmid, and BAC engineering, plus homologous recombination.
  • Enables precise modification using galK positive/negative selection.
  • Derived from SW102 (10B background).
  • Heat-inducible prophage with red genes (exo, bet, gam), repressed at 32 °C and induced at 42 °C.
  • Tetracycline resistant (TetR: 5 μg/ml).