Learn More
INTACT GENOMICS INC ig 10B Chemically Competent Cells (High Efficiency ), 1x384 well plate - 15µl/well
Catalog No. NC3998220
Encompass
These chemically competent E. coli cells are designed for high-efficiency transformation in various applications like cloning and sub-cloning. They are a derivative of DH10B and offer industry-leading transformation efficiency. This product is provided as a 1x384 well plate.
  • Φ80lacZΔM15 marker provides α-complementation of the β-galactosidase gene with blue/white screening.
  • McrA genotypic marker and the mcrBC, mrr deletion allow for cloning DNA that contains methylcytosine and methyladenine.
  • Offers the highest transformation efficiency on the market.
  • Ideal for creating cDNA or genomic libraries.
  • Suitable for transforming large plasmids (>10 kb).
  • Transformation efficiency of ≥1.0 x 1010 cfu/μg pUC19 DNA.
  • Blue/white screening capability.
  • Provided in a 384-well format (10μL/well).
Catalog No. NC3998220 Supplier INTACT GENOMICS INC Supplier No. 1011384
These chemically competent E. coli cells are designed for high-efficiency transformation in various applications like cloning and sub-cloning. They are a derivative of DH10B and offer industry-leading transformation efficiency. This product is provided as a 1x384 well plate.
  • Φ80lacZΔM15 marker provides α-complementation of the β-galactosidase gene with blue/white screening.
  • McrA genotypic marker and the mcrBC, mrr deletion allow for cloning DNA that contains methylcytosine and methyladenine.
  • Offers the highest transformation efficiency on the market.
  • Ideal for creating cDNA or genomic libraries.
  • Suitable for transforming large plasmids (>10 kb).
  • Transformation efficiency of ≥1.0 x 1010 cfu/μg pUC19 DNA.
  • Blue/white screening capability.
  • Provided in a 384-well format (10μL/well).