Learn More
INTACT GENOMICS INC IG Autoinduction DE3 Chemically Competent Cells, 24x50µl
Catalog No. NC3998322
Encompass
Intact Genomics BL21(DE3) Chemically Competent E. coli Cells are suitable for transformation and routine protein expression.
BL21(DE3) Chemically Competent Cells have the following features:
Widely used host background
T7 Expression Strain
Deficient in both lon (1) and ompT proteases
Resistant to phage T1 (fhuA2)
B Strain
Product Includes and Storage:
BL21(DE3) Competent Cells: -80 ºC
pUC19 Control DNA: -20 ºC
Recovery Medium: 4 ºC
Catalog No. NC3998322 Supplier INTACT GENOMICS INC Supplier No. 106524
Intact Genomics BL21(DE3) Chemically Competent E. coli Cells are suitable for transformation and routine protein expression.
BL21(DE3) Chemically Competent Cells have the following features:
Widely used host background
T7 Expression Strain
Deficient in both lon (1) and ompT proteases
Resistant to phage T1 (fhuA2)
B Strain
Product Includes and Storage:
BL21(DE3) Competent Cells: -80 ºC
pUC19 Control DNA: -20 ºC
Recovery Medium: 4 ºC