Learn More
INTACT GENOMICS INC IG C41(DE3) Chemically Competent Cells 6x50µl
Catalog No. NC3998268
Encompass
C41(DE3) Chemically Competent Cells are engineered for transformation and recombinant protein expression using IPTG-inducible T7 promoters. Derived from a BL21(DE3) mutant, this strain prevents cell death commonly associated with expressing toxic recombinant proteins, offering a solution when standard BL21(DE3) strains are insufficient. These cells efficiently express toxic proteins from a diverse range of organisms, validated by extensive research.
  • Engineered for improved tolerance to toxic protein expression.
  • Maintains reliable T7-driven performance.
  • Derived from BL21(DE3) to reduce cell death.
  • Suitable for expression of toxic proteins from diverse organisms.
  • Transformation efficiency of at least 3.0 x 107 cfu/μg pUC19 DNA.
  • Chemically competent cell type.
  • Species: *E. coli*.
  • Strain: C41(DE3).
  • Supplied in tubes.
  • Shipped on dry ice.
Catalog No. NC3998268 Supplier INTACT GENOMICS INC Supplier No. 104306
C41(DE3) Chemically Competent Cells are engineered for transformation and recombinant protein expression using IPTG-inducible T7 promoters. Derived from a BL21(DE3) mutant, this strain prevents cell death commonly associated with expressing toxic recombinant proteins, offering a solution when standard BL21(DE3) strains are insufficient. These cells efficiently express toxic proteins from a diverse range of organisms, validated by extensive research.
  • Engineered for improved tolerance to toxic protein expression.
  • Maintains reliable T7-driven performance.
  • Derived from BL21(DE3) to reduce cell death.
  • Suitable for expression of toxic proteins from diverse organisms.
  • Transformation efficiency of at least 3.0 x 107 cfu/μg pUC19 DNA.
  • Chemically competent cell type.
  • Species: *E. coli*.
  • Strain: C41(DE3).
  • Supplied in tubes.
  • Shipped on dry ice.