Learn More
INTACT GENOMICS INC DirectPlate DH5-alpha Chemically Competent Cells , 1x384 well plate - 20µl/well
Catalog No. NC3998222
Encompass
Intact Genomics DirectPlate™ Competent cells offer simple, fast, and robust DNA transformation. DirectPlate™ DH5-Alpha chemically competent E. coli cells streamline the workflow by eliminating heat shock, lengthy incubations, and time-consuming outgrowth procedures. Users simply mix and directly plate. These cells provide higher transformation efficiency than similar competitor products and are suitable for high-efficiency cloning and sub-cloning applications.
  • Simple, fast, and robust results for DNA transformation
  • Eliminates heat shock
  • Eliminates lengthy incubations
  • Eliminates time-consuming outgrowth procedures
  • Simply mix and directly plate
  • Higher transformation efficiency than competitor’s similar products
  • Suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning
Catalog No. NC3998222 Supplier INTACT GENOMICS INC Supplier No. 1013384
Intact Genomics DirectPlate™ Competent cells offer simple, fast, and robust DNA transformation. DirectPlate™ DH5-Alpha chemically competent E. coli cells streamline the workflow by eliminating heat shock, lengthy incubations, and time-consuming outgrowth procedures. Users simply mix and directly plate. These cells provide higher transformation efficiency than similar competitor products and are suitable for high-efficiency cloning and sub-cloning applications.
  • Simple, fast, and robust results for DNA transformation
  • Eliminates heat shock
  • Eliminates lengthy incubations
  • Eliminates time-consuming outgrowth procedures
  • Simply mix and directly plate
  • Higher transformation efficiency than competitor’s similar products
  • Suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning