Learn More
INTACT GENOMICS INC IG C41(DE3) Electro Competent Cells 24x50μl
Catalog No. NC3998370
Encompass
Intact Genomics C41(DE3) Electrocompetent Cells are engineered for improved tolerance to the expression of toxic proteins. Derived from the BL21(DE3) strain, these E. coli cells minimize cell death during challenging recombinant protein expression. They are suitable for transformation and routine protein expression driven by an IPTG-inducible T7 promoter, enabling efficient expression of toxic genes from diverse organisms.
  • Engineered for improved tolerance to toxic protein expression.
  • Minimizes cell death for membrane proteins and nucleases.
  • Designed for routine recombinant protein expression with IPTG-inducible T7 promoter.
  • Enables efficient expression of toxic genes from various organisms.
  • T7 expression strain.
  • Transformation efficiency ≥ 1.0 x 1010 cfu/μg pUC19 DNA.
Catalog No. NC3998370 Supplier INTACT GENOMICS INC Supplier No. 124348
Intact Genomics C41(DE3) Electrocompetent Cells are engineered for improved tolerance to the expression of toxic proteins. Derived from the BL21(DE3) strain, these E. coli cells minimize cell death during challenging recombinant protein expression. They are suitable for transformation and routine protein expression driven by an IPTG-inducible T7 promoter, enabling efficient expression of toxic genes from diverse organisms.
  • Engineered for improved tolerance to toxic protein expression.
  • Minimizes cell death for membrane proteins and nucleases.
  • Designed for routine recombinant protein expression with IPTG-inducible T7 promoter.
  • Enables efficient expression of toxic genes from various organisms.
  • T7 expression strain.
  • Transformation efficiency ≥ 1.0 x 1010 cfu/μg pUC19 DNA.