Learn More
INTACT GENOMICS INC igMa 10B ElectroCompetent Cells, 12x100l
Catalog No. NC3998420
Encompass
Intact Genomics igMax™ 10B derivative electrocompetent cells are engineered for superior transformation of large DNA molecules (≥10 kb up to 350 kb). They deliver exceptional performance with transformation efficiencies of ≥5 × 10¹&sup0; cfu/μg DNA, making them ideal for cloning large plasmids and BACs in demanding applications.
  • Engineered for superior transformation of large DNA molecules (≥10 kb up to 350 kb).
  • Exceptional performance with transformation efficiencies of ≥5 × 10¹&sup0; cfu/μg DNA with electroporation.
  • Ideal for cloning large plasmids, BACs, and high-molecular-weight constructs.
  • Suitable for demanding cloning applications such as synthetic bio-applications and assembling large multi-DNA fragments.
  • Utilizes proprietary manufacturing methods for effective transformation of all large DNA molecules.
Catalog No. NC3998420 Supplier INTACT GENOMICS INC Supplier No. 128448
Intact Genomics igMax™ 10B derivative electrocompetent cells are engineered for superior transformation of large DNA molecules (≥10 kb up to 350 kb). They deliver exceptional performance with transformation efficiencies of ≥5 × 10¹&sup0; cfu/μg DNA, making them ideal for cloning large plasmids and BACs in demanding applications.
  • Engineered for superior transformation of large DNA molecules (≥10 kb up to 350 kb).
  • Exceptional performance with transformation efficiencies of ≥5 × 10¹&sup0; cfu/μg DNA with electroporation.
  • Ideal for cloning large plasmids, BACs, and high-molecular-weight constructs.
  • Suitable for demanding cloning applications such as synthetic bio-applications and assembling large multi-DNA fragments.
  • Utilizes proprietary manufacturing methods for effective transformation of all large DNA molecules.