Learn More
INTACT GENOMICS INC IG C43(DE3)pLysS Electro Competent Cells 6x50µl
Catalog No. NC3998395
Encompass
These electrocompetent cells are for high-efficiency transformation and stable expression of toxic or difficult proteins in IPTG-inducible T7 systems. Derived from BL21(DE3), they offer improved tolerance to toxic proteins. The pLysS plasmid suppresses basal expression, stabilizing toxic recombinant proteins.
  • Reliable expression for toxic proteins.
  • High-efficiency transformation.
  • Derived from BL21(DE3); improved tolerance to toxic proteins.
  • pLysS plasmid suppresses basal expression for challenging constructs.
  • Effective for expressing toxic proteins from diverse organisms.
  • Contains T7 lysozyme to suppress basal expression.
  • Validated in over 400 publications.
  • T7 expression strain.
  • Transformation efficiency: ≥ 5.0 x 109 cfu/μg pUC19 DNA.
  • Shipped on dry ice.
  • Store cells at –80°C.
Catalog No. NC3998395 Supplier INTACT GENOMICS INC Supplier No. 124912
These electrocompetent cells are for high-efficiency transformation and stable expression of toxic or difficult proteins in IPTG-inducible T7 systems. Derived from BL21(DE3), they offer improved tolerance to toxic proteins. The pLysS plasmid suppresses basal expression, stabilizing toxic recombinant proteins.
  • Reliable expression for toxic proteins.
  • High-efficiency transformation.
  • Derived from BL21(DE3); improved tolerance to toxic proteins.
  • pLysS plasmid suppresses basal expression for challenging constructs.
  • Effective for expressing toxic proteins from diverse organisms.
  • Contains T7 lysozyme to suppress basal expression.
  • Validated in over 400 publications.
  • T7 expression strain.
  • Transformation efficiency: ≥ 5.0 x 109 cfu/μg pUC19 DNA.
  • Shipped on dry ice.
  • Store cells at –80°C.