Learn More
INTACT GENOMICS INC IG Autoinduction DE3 ElectroCompetent Cells, 6x50µl
Catalog No. NC3998405
Encompass
These Autoinduction DE3 ElectroCompetent cells are designed for efficient recombinant protein expression. Featuring a chromosomally integrated IPTG-inducible promoter for T7 RNA polymerase, this strain simplifies the process and significantly increases protein yields for various targets.
  • Efficient recombinant protein expression with IPTG-inducible T7 promoter.
  • Simplified autoinduction.
  • T7 expression strain, optimized for autoinduction.
  • Suitable for expressing toxic genes.
  • Tetracycline resistant (12.5 μg/mL).
  • Electroporation competent E. coli.
  • Transformation efficiency: ≥ 1.0 x 1010 cfu/μg pUC19 DNA.
  • Supplied in tubes; shipped on dry ice.
  • Storage: Cells -80 °C, pUC19 DNA -20 °C, recovery medium 4 °C.
Catalog No. NC3998405 Supplier INTACT GENOMICS INC Supplier No. 126512
These Autoinduction DE3 ElectroCompetent cells are designed for efficient recombinant protein expression. Featuring a chromosomally integrated IPTG-inducible promoter for T7 RNA polymerase, this strain simplifies the process and significantly increases protein yields for various targets.
  • Efficient recombinant protein expression with IPTG-inducible T7 promoter.
  • Simplified autoinduction.
  • T7 expression strain, optimized for autoinduction.
  • Suitable for expressing toxic genes.
  • Tetracycline resistant (12.5 μg/mL).
  • Electroporation competent E. coli.
  • Transformation efficiency: ≥ 1.0 x 1010 cfu/μg pUC19 DNA.
  • Supplied in tubes; shipped on dry ice.
  • Storage: Cells -80 °C, pUC19 DNA -20 °C, recovery medium 4 °C.