Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INTACT GENOMICS INC Staphylococcus aureus (S. aureus) RN4220 Electrocompetent Cells 10x50µl

Catalog No. NC3998430
Encompass

Intact Genomics (ig®) Staphylococcus aureus (S. aureus) RN4220 Electrocompetent Cells offer high transformation efficiencies, suitable for studies involving virulence, resistance, and metabolism. This RN4220 strain is restriction deficient due to a mutation in the sau1 hsdR genes, making it an effective intermediate cloning host.

  • High transformation efficiencies for cloning and various applications.
  • Restriction deficient (sau1 hsdR gene mutation).
  • Competent cell type: Electrocompetent.
  • Species: S. aureus.
  • Strain: RN4220.
  • Format: Tubes.
  • Transformation efficiency: ≥1 x 105 cfu/μg DNA.
  • No blue/white screening.
  • Shipping condition: Dry ice.
  • Storage: -80 °C for cells, 4 °C for recovery medium.
  • Biosafety Level 2.

Catalog No. NC3998430 Supplier INTACT GENOMICS INC Supplier No. 129420
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Intact Genomics (ig®) Staphylococcus aureus (S. aureus) RN4220 Electrocompetent Cells offer high transformation efficiencies, suitable for studies involving virulence, resistance, and metabolism. This RN4220 strain is restriction deficient due to a mutation in the sau1 hsdR genes, making it an effective intermediate cloning host.

  • High transformation efficiencies for cloning and various applications.
  • Restriction deficient (sau1 hsdR gene mutation).
  • Competent cell type: Electrocompetent.
  • Species: S. aureus.
  • Strain: RN4220.
  • Format: Tubes.
  • Transformation efficiency: ≥1 x 105 cfu/μg DNA.
  • No blue/white screening.
  • Shipping condition: Dry ice.
  • Storage: -80 °C for cells, 4 °C for recovery medium.
  • Biosafety Level 2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.