Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INTACT GENOMICS INC IG C41(DE3)pLysS Chemically Competent Cells 6x50µl

Catalog No. NC3998282
Encompass

Chemically competent E. coli cells are designed for high efficiency transformation in various applications, including cloning and sub-cloning.

  • Suitable for high efficiency transformation
  • Ideal for cloning and sub-cloning applications
  • Competent cell type: Chemically competent
  • Derivative of DH5a
  • Species: E. coli
  • Format: Tubes
  • Transformation efficiency: ≥1 x 109 cfu/μg pUC19 DNA
  • Blue/white screening available
  • Shipped on dry ice
  • Storage at -80 °C

Catalog No. NC3998282 Supplier INTACT GENOMICS INC Supplier No. 104506
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemically competent E. coli cells are designed for high efficiency transformation in various applications, including cloning and sub-cloning.

  • Suitable for high efficiency transformation
  • Ideal for cloning and sub-cloning applications
  • Competent cell type: Chemically competent
  • Derivative of DH5a
  • Species: E. coli
  • Format: Tubes
  • Transformation efficiency: ≥1 x 109 cfu/μg pUC19 DNA
  • Blue/white screening available
  • Shipped on dry ice
  • Storage at -80 °C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.