Learn More
INTACT GENOMICS INC BL21(DE3) Chemically Competent Cells , 96x50µl (12x8-well strip)
Catalog No. NC3998314
Encompass
These BL21(DE3) Chemically Competent E. coli Cells offer high transformation efficiency and reliable performance for cloning and recombinant protein production. They are optimized for T7-based expression systems and are suitable for transformation and routine protein expression.
  • High transformation efficiency
  • Excellent performance for protein expression
  • Custom formats available: tubes, 96-well plates, and 384-well plates
  • Flexible solutions for high-throughput workflows
  • Widely used host background
  • T7 expression strain
  • Deficient in both lon and ompT proteases
  • Resistant to phage T1 (fhuA2)
  • B strain
Catalog No. NC3998314 Supplier INTACT GENOMICS INC Supplier No. 105595
These BL21(DE3) Chemically Competent E. coli Cells offer high transformation efficiency and reliable performance for cloning and recombinant protein production. They are optimized for T7-based expression systems and are suitable for transformation and routine protein expression.
  • High transformation efficiency
  • Excellent performance for protein expression
  • Custom formats available: tubes, 96-well plates, and 384-well plates
  • Flexible solutions for high-throughput workflows
  • Widely used host background
  • T7 expression strain
  • Deficient in both lon and ompT proteases
  • Resistant to phage T1 (fhuA2)
  • B strain