Learn More
INTACT GENOMICS INC IG C41(DE3) Electro Competent Cells 6x50μl
Supplier: INTACT GENOMICS INC 124312
These electrocompetent cells are engineered to improve tolerance to toxic protein expression while maintaining reliable T7-driven performance. Derived from BL21(DE3), the C41(DE3) strain reduces cell death associated with membrane proteins, nucleases, and other challenging targets. These cells are designed for transformation and routine recombinant protein expression from vectors under an IPTG-inducible T7 promoter.
- T7 expression strain
- Selected for expression of toxic proteins
- Suitable for expression of toxic genes
- Electrocompetent cell type
- Species: E. coli
- Strain: C41(DE3)
- Format: Tubes
- Transformation efficiency: ≥ 1.0 x 1010 cfu/μg pUC19 DNA
- Blue/white screening: No
- Shipping condition: Dry ice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.