Learn More
INTACT GENOMICS INC IG C43(DE3) Electro Competent Cells 24x50µl
Catalog No. NC3998385
Encompass
These C43(DE3)pLysS Electrocompetent Cells are optimized for the expression of toxic and difficult-to-express proteins from vectors with an IPTG-inducible T7 promoter. They are a mutant derivative of BL21(DE3) and contain additional mutations to help prevent cell death associated with the expression of many recombinant toxic proteins. This strain is effective for expressing toxic proteins from a wide range of organisms.
  • Electrocompetent cell type
  • E. coli species
  • C43(DE3) strain
  • Supplied in tubes
  • Transformation efficiency ≥ 1.0 x 1010 CFU/μg pUC19 DNA
  • No blue/white screening
  • Shipped on dry ice
  • T7 expression strain
  • Selected for expression of toxic proteins
  • Suitable for expression of toxic genes
Catalog No. NC3998385 Supplier INTACT GENOMICS INC Supplier No. 124748
These C43(DE3)pLysS Electrocompetent Cells are optimized for the expression of toxic and difficult-to-express proteins from vectors with an IPTG-inducible T7 promoter. They are a mutant derivative of BL21(DE3) and contain additional mutations to help prevent cell death associated with the expression of many recombinant toxic proteins. This strain is effective for expressing toxic proteins from a wide range of organisms.
  • Electrocompetent cell type
  • E. coli species
  • C43(DE3) strain
  • Supplied in tubes
  • Transformation efficiency ≥ 1.0 x 1010 CFU/μg pUC19 DNA
  • No blue/white screening
  • Shipped on dry ice
  • T7 expression strain
  • Selected for expression of toxic proteins
  • Suitable for expression of toxic genes