Learn More
INTACT GENOMICS INC BL21(DE3) Chemically Competent Cells , 1x96 well plate (20µl/well)
Catalog No. NC3998316
Encompass
Intact Genomics (IG™) BL21(DE3) Chemically Competent Cells offer high transformation efficiency and reliable performance for cloning and recombinant protein production. They are optimized for T7-based expression systems and provide consistent results. Intact Genomics chemically competent BL21(DE3) E. coli cells are suitable for transformation and routine protein expression.
  • High transformation efficiency
  • Excellent performance for protein expression
  • Custom formats available: tubes, 96-well plates, and 384-well plates
  • Flexible solutions for high-throughput workflows
  • Widely used host background
  • T7 Expression Strain
  • Deficient in both lon and ompT proteases
  • Resistant to phage T1 (fhuA2)
  • B Strain
Catalog No. NC3998316 Supplier INTACT GENOMICS INC Supplier No. 105596
Intact Genomics (IG™) BL21(DE3) Chemically Competent Cells offer high transformation efficiency and reliable performance for cloning and recombinant protein production. They are optimized for T7-based expression systems and provide consistent results. Intact Genomics chemically competent BL21(DE3) E. coli cells are suitable for transformation and routine protein expression.
  • High transformation efficiency
  • Excellent performance for protein expression
  • Custom formats available: tubes, 96-well plates, and 384-well plates
  • Flexible solutions for high-throughput workflows
  • Widely used host background
  • T7 Expression Strain
  • Deficient in both lon and ompT proteases
  • Resistant to phage T1 (fhuA2)
  • B Strain