Learn More
INTACT GENOMICS INC IG Autoinduction DE3 ElectroCompetent Cells, 24x50µl
Catalog No. NC3998413
Encompass
IG™ Autoinduction DE3 ElectroCompetent cells are suitable for transformation and routine recombinant protein expression from an IPTG-inducible T7 promoter vector. This Autoinduction strain features a chromosomally integrated IPTG-inducible promoter, enabling T7 RNA polymerase expression via IPTG or optimized autoinduction media. They offer simple, less hands-on autoinduction, potentially boosting yields of expressed cells and protein 5 to 10 fold for various protein targets.
  • Suitable for transformation and routine recombinant protein expression from an IPTG-inducible T7 promoter vector.
  • Simple, less hands-on autoinduction.
  • Yields of expressed cells and protein are likely 5 to 10 fold or more for a broad range of protein targets.
  • T7 expression strain optimized for autoinduction.
  • Suitable for expression of toxic genes.
  • Tetracycline resistant (12.5 μg/mL).
Catalog No. NC3998413 Supplier INTACT GENOMICS INC Supplier No. 126548
IG™ Autoinduction DE3 ElectroCompetent cells are suitable for transformation and routine recombinant protein expression from an IPTG-inducible T7 promoter vector. This Autoinduction strain features a chromosomally integrated IPTG-inducible promoter, enabling T7 RNA polymerase expression via IPTG or optimized autoinduction media. They offer simple, less hands-on autoinduction, potentially boosting yields of expressed cells and protein 5 to 10 fold for various protein targets.
  • Suitable for transformation and routine recombinant protein expression from an IPTG-inducible T7 promoter vector.
  • Simple, less hands-on autoinduction.
  • Yields of expressed cells and protein are likely 5 to 10 fold or more for a broad range of protein targets.
  • T7 expression strain optimized for autoinduction.
  • Suitable for expression of toxic genes.
  • Tetracycline resistant (12.5 μg/mL).