Learn More
INTACT GENOMICS INC IG C41(DE3)pLysS Electro Competent Cells 6x50µl
Catalog No. NC3998372
Encompass
IG C41(DE3)pLysS Electrocompetent Cells are designed for transformation and recombinant protein expression using IPTG-inducible T7 promoter vectors. Derived from BL21(DE3), these cells improve tolerance to toxic proteins, preventing cell death during expression. They contain a pLysS plasmid with T7 lysozyme, suppressing basal T7 RNA polymerase expression and stabilizing recombinants with highly toxic proteins. Its efficacy is validated in over 400 publications.
  • Built for tough protein expression.
  • Designed for IPTG-inducible T7 systems.
  • Improves tolerance to toxic proteins.
  • PlysS plasmid suppresses basal expression for greater stability.
  • Validated in over 400 publications.
  • T7 expression strain.
  • Selected for expression of toxic proteins.
  • Suitable for expression of toxic genes.
Catalog No. NC3998372 Supplier INTACT GENOMICS INC Supplier No. 124512
IG C41(DE3)pLysS Electrocompetent Cells are designed for transformation and recombinant protein expression using IPTG-inducible T7 promoter vectors. Derived from BL21(DE3), these cells improve tolerance to toxic proteins, preventing cell death during expression. They contain a pLysS plasmid with T7 lysozyme, suppressing basal T7 RNA polymerase expression and stabilizing recombinants with highly toxic proteins. Its efficacy is validated in over 400 publications.
  • Built for tough protein expression.
  • Designed for IPTG-inducible T7 systems.
  • Improves tolerance to toxic proteins.
  • PlysS plasmid suppresses basal expression for greater stability.
  • Validated in over 400 publications.
  • T7 expression strain.
  • Selected for expression of toxic proteins.
  • Suitable for expression of toxic genes.