Learn More
INTACT GENOMICS INC IG C43(DE3)pLysS Electro Competent Cells 24x50µl
Catalog No. NC3998399
Encompass
IG™ C43(DE3)pLysS Electrocompetent Cells are designed for high-efficiency transformation and stable expression of toxic or difficult proteins in IPTG-inducible T7 systems. This BL21(DE3) derived strain includes mutations improving tolerance to toxic proteins. It carries a chloramphenicol-resistant pLysS plasmid encoding T7 lysozyme, suppressing basal T7 RNA polymerase expression to stabilize recombinants. Effectiveness is validated in over 400 publications.
  • T7 expression strain
  • Designed for expression of toxic proteins
  • BL21(DE3) derived with improved tolerance mutations
  • Contains chloramphenicol-resistant pLysS plasmid
  • Suppresses basal T7 RNA polymerase expression
  • Validated in over 400 publications
Catalog No. NC3998399 Supplier INTACT GENOMICS INC Supplier No. 124948
IG™ C43(DE3)pLysS Electrocompetent Cells are designed for high-efficiency transformation and stable expression of toxic or difficult proteins in IPTG-inducible T7 systems. This BL21(DE3) derived strain includes mutations improving tolerance to toxic proteins. It carries a chloramphenicol-resistant pLysS plasmid encoding T7 lysozyme, suppressing basal T7 RNA polymerase expression to stabilize recombinants. Effectiveness is validated in over 400 publications.
  • T7 expression strain
  • Designed for expression of toxic proteins
  • BL21(DE3) derived with improved tolerance mutations
  • Contains chloramphenicol-resistant pLysS plasmid
  • Suppresses basal T7 RNA polymerase expression
  • Validated in over 400 publications