Learn More
INTACT GENOMICS INC IG C43(DE3) Electro Competent Cells 6x50µl
Supplier:  INTACT GENOMICS INC 124712
Encompass
Intact Genomics C43(DE3) Electrocompetent Cells are optimized for the expression of toxic and difficult proteins from vectors with an IPTG-inducible T7 promoter. These mutant BL21(DE3) derivative cells carry mutations preventing cell death during expression of many recombinant toxic proteins, making them highly effective for toxic protein expression from diverse organisms.
  • T7 expression strain for toxic proteins.
  • Electrocompetent E. coli cells, strain C43(DE3).
  • Supplied in tubes.
  • Transformation efficiency: ≥ 1.0 x 1010 cfu/μg pUC19 DNA.
  • No blue/white screening.
  • Shipped on dry ice.
  • Mutant BL21(DE3) derivative with mutations preventing cell death during recombinant toxic protein expression.
Catalog No. NC3998381