Learn More
INTACT GENOMICS INC ig ER2738 Phage Display ElectroCompetent Cells, 6x50µl
Catalog No. NC3998350
Encompass
This product is an electrocompetent competent cell specifically designed for Phage Display applications.
  • Volume: 6x50 μl
  • Efficiency: ≥4.0 x 1010
Catalog No. NC3998350 Supplier INTACT GENOMICS INC Supplier No. 120712
This product is an electrocompetent competent cell specifically designed for Phage Display applications.
  • Volume: 6x50 μl
  • Efficiency: ≥4.0 x 1010