Learn More
INTACT GENOMICS INC IG RPA Master Mix, 500 Reactions 
Catalog No. NC3998206
Encompass
IG RPA Master Mix is a ready to use premix which contains T4 UvsX, UvxY, gp32, Bsu DNA polymerase, and stabilizers with an optimized reaction solution. It simplifies and quickens the setup for your RPA reaction. This patent pending, proprietary IG RPA Master Mix offers two key advantages over other RPA products: 1) More robust amplification at shorter times and 2) Lower background amplification of off-target products meaning IG RPA Master Mix has more specific amplification of the primary target.
Catalog No. NC3998206 Supplier INTACT GENOMICS INC Supplier No. 3549
IG RPA Master Mix is a ready to use premix which contains T4 UvsX, UvxY, gp32, Bsu DNA polymerase, and stabilizers with an optimized reaction solution. It simplifies and quickens the setup for your RPA reaction. This patent pending, proprietary IG RPA Master Mix offers two key advantages over other RPA products: 1) More robust amplification at shorter times and 2) Lower background amplification of off-target products meaning IG RPA Master Mix has more specific amplification of the primary target.