Learn More
INTACT GENOMICS INC IG RPA master mix, Lyophilized, 500 Reactions
Catalog No. NC3998444
Encompass
This RPA Master Mix is a ready-to-use premix designed for Recombinase Polymerase Amplification (RPA) reactions. It offers robust, specific isothermal amplification for molecular diagnostics and research, working efficiently at 25°C to 37°C without a thermal cycler.
  • Works efficiently at 25°C to 37°C, eliminating the need for a thermal cycler.
  • Simplifies RPA reaction setup and sample preparation, requiring no DNA pretreatment.
  • Provides robust, specific, high-sensitivity amplification with consistent results and low background.
  • Rapid amplification: DNA in 10 minutes at 25°C, or 5 minutes at 37°C.
  • Suitable for various diagnostic and monitoring applications; compatible with lateral flow and real-time fluorescence.
Catalog No. NC3998444 Supplier INTACT GENOMICS INC Supplier No. 3549LYO
This RPA Master Mix is a ready-to-use premix designed for Recombinase Polymerase Amplification (RPA) reactions. It offers robust, specific isothermal amplification for molecular diagnostics and research, working efficiently at 25°C to 37°C without a thermal cycler.
  • Works efficiently at 25°C to 37°C, eliminating the need for a thermal cycler.
  • Simplifies RPA reaction setup and sample preparation, requiring no DNA pretreatment.
  • Provides robust, specific, high-sensitivity amplification with consistent results and low background.
  • Rapid amplification: DNA in 10 minutes at 25°C, or 5 minutes at 37°C.
  • Suitable for various diagnostic and monitoring applications; compatible with lateral flow and real-time fluorescence.