Learn More
INTACT GENOMICS INC Recombineering Chemically Competent Cells 24x50l
Catalog No. NC3998346
Encompass
Intact Genomics Recombineering Chemically Competent Cells are designed for high-efficiency transformation in DNA, plasmid, and BAC engineering, as well as homologous recombination. This SW102 derivative strain facilitates efficient DNA or BAC modification through galK positive/negative selection and is tetracycline resistant (5 μg/mL).
  • Chemically competent cell type.
  • E. coli species.
  • Provided in tubes.
  • Transformation efficiency: ≥ 1.0 x 109 cfu/μg pUC19 DNA.
  • Features red genes (exo, bet, gam) under heat-inducible promoter pL, repressed at 32 °C and induced at 42 °C.
  • ΔgalK derivative supports galK positive/negative selection.
  • Tetracycline resistant (TetR) via a gene replacement (5 μg/mL).
  • Genotype: 10B [λcI857ind1 (cro-bioA<>tet)].
  • Ships on dry ice.
  • Available in 24x50 μL quantity.
Catalog No. NC3998346 Supplier INTACT GENOMICS INC Supplier No. 106624
Intact Genomics Recombineering Chemically Competent Cells are designed for high-efficiency transformation in DNA, plasmid, and BAC engineering, as well as homologous recombination. This SW102 derivative strain facilitates efficient DNA or BAC modification through galK positive/negative selection and is tetracycline resistant (5 μg/mL).
  • Chemically competent cell type.
  • E. coli species.
  • Provided in tubes.
  • Transformation efficiency: ≥ 1.0 x 109 cfu/μg pUC19 DNA.
  • Features red genes (exo, bet, gam) under heat-inducible promoter pL, repressed at 32 °C and induced at 42 °C.
  • ΔgalK derivative supports galK positive/negative selection.
  • Tetracycline resistant (TetR) via a gene replacement (5 μg/mL).
  • Genotype: 10B [λcI857ind1 (cro-bioA<>tet)].
  • Ships on dry ice.
  • Available in 24x50 μL quantity.