Learn More
INTACT GENOMICS INC Recovery Media, 100mL
Catalog No. NC3998088
Encompass
Intact Genomics Recovery Medium is a superiorly formulated solution recommended for all competent cell transformations. It is designed to maximize transformation efficiency with any competent cell type. The medium features an enhanced SOC formulation and is provided with all Intact Genomics competent cells.
  • Superior formulation for competent cell transformations.
  • Achieves the highest possible transformation efficiency from any competent cell.
  • Enhanced SOC formulation.
  • Shipped frozen but should be stored at 4°C upon receipt.
Catalog No. NC3998088 Supplier INTACT GENOMICS INC Supplier No. 1715
Intact Genomics Recovery Medium is a superiorly formulated solution recommended for all competent cell transformations. It is designed to maximize transformation efficiency with any competent cell type. The medium features an enhanced SOC formulation and is provided with all Intact Genomics competent cells.
  • Superior formulation for competent cell transformations.
  • Achieves the highest possible transformation efficiency from any competent cell.
  • Enhanced SOC formulation.
  • Shipped frozen but should be stored at 4°C upon receipt.