Learn More
INTACT GENOMICS INC Recovery Media, 4x30mL
Catalog No. NC3998076
Encompass
Intact Genomics (ig™) Recovery/Outgrowth Medium is a specialized solution with a superior formulation, recommended for all competent cell transformations.
  • Recommended for all competent cell transformations.
  • Formulated to achieve the highest possible transformation efficiency from any competent cell.
  • Features an enhanced SOC formulation.
  • Package size: 8 x 12 mL.
  • Shipped frozen; store at 4 °C upon receipt.
Catalog No. NC3998076 Supplier INTACT GENOMICS INC Supplier No. 1712
Intact Genomics (ig™) Recovery/Outgrowth Medium is a specialized solution with a superior formulation, recommended for all competent cell transformations.
  • Recommended for all competent cell transformations.
  • Formulated to achieve the highest possible transformation efficiency from any competent cell.
  • Features an enhanced SOC formulation.
  • Package size: 8 x 12 mL.
  • Shipped frozen; store at 4 °C upon receipt.