Learn More
INTACT GENOMICS INC IG RPA Master Mix,  100 Reactions 
Supplier:  INTACT GENOMICS INC 3547
Encompass
This RPA Master Mix is a ready-to-use premix, containing T4 UvsX, UvsY, gp32, Bsu DNA polymerase, and stabilizers in an optimized reaction solution. It simplifies and accelerates RPA reaction setup, providing robust amplification and reducing background for specific primary target amplification. It delivers powerful isothermal amplification for molecular diagnostics and research.
  • Operates at 25–37°C, eliminating the need for a thermal cycler.
  • Optimized for high sensitivity, consistent results, and simplified workflow.
  • Provides robust and specific amplification in shorter times.
  • Highly selective and sensitive isothermal amplification technique.
  • Excellent for rapid point-of-care and on-site testing.
  • Compatible with flexible endpoint detection methods.
Catalog No. NC3998193