Learn More
INTACT GENOMICS INC IG RPA Master Mix, 25 Reactions
Catalog No. NC3998181
Encompass
This RPA Master Mix is a ready-to-use premix with T4 UvsX, UvsY, gp32, Bsu DNA polymerase, and stabilizers for optimized reaction setup. It enables robust, fast amplification with lower background for specific primary target amplification.
  • Works at 25–37°C, no thermal cycler needed.
  • Affordable, accessible, and optimized for high sensitivity.
  • Simplifies workflow and amplifies outcomes.
  • Premix includes T4 UvsX, UvsY, gp32, Bsu DNA polymerase, and stabilizers.
  • Achieves robust, faster amplification with lower off-target background.
  • Highly selective and sensitive isothermal amplification.
  • Delivers speed for rapid molecular and agricultural tests.
  • Compatible with flexible endpoint detection (e.g., lateral flow).
  • Excellent for rapid point-of-care and on-site testing.
Catalog No. NC3998181 Supplier INTACT GENOMICS INC Supplier No. 3545
This RPA Master Mix is a ready-to-use premix with T4 UvsX, UvsY, gp32, Bsu DNA polymerase, and stabilizers for optimized reaction setup. It enables robust, fast amplification with lower background for specific primary target amplification.
  • Works at 25–37°C, no thermal cycler needed.
  • Affordable, accessible, and optimized for high sensitivity.
  • Simplifies workflow and amplifies outcomes.
  • Premix includes T4 UvsX, UvsY, gp32, Bsu DNA polymerase, and stabilizers.
  • Achieves robust, faster amplification with lower off-target background.
  • Highly selective and sensitive isothermal amplification.
  • Delivers speed for rapid molecular and agricultural tests.
  • Compatible with flexible endpoint detection (e.g., lateral flow).
  • Excellent for rapid point-of-care and on-site testing.