Learn More
INTACT GENOMICS INC IG RPA master mix, Lyophilized, 25 Reactions
Catalog No. NC3998439
Encompass
The IG® RPA Master Mix is a fast, reliable, isothermal amplification solution for molecular diagnostics and research. It offers high sensitivity and consistent results, operating at 25–37°C without a thermal cycler. This ready-to-use premix simplifies setup and ensures robust, specific amplification.
  • Isothermal amplification: 25–37°C operation, no thermal cycler needed.
  • Ready-to-use premix for simplified reaction setup.
  • Enhanced specificity: lower background, more specific target DNA amplification.
  • Rapid amplification: DNA amplifiable in 5–10 min.
  • Cost-effective and accessible for diagnostics and research.
  • Flexible formats: liquid and lyophilized options available.
  • Diverse applications in healthcare, food safety, environmental monitoring.
Catalog No. NC3998439 Supplier INTACT GENOMICS INC Supplier No. 3545LYO
The IG® RPA Master Mix is a fast, reliable, isothermal amplification solution for molecular diagnostics and research. It offers high sensitivity and consistent results, operating at 25–37°C without a thermal cycler. This ready-to-use premix simplifies setup and ensures robust, specific amplification.
  • Isothermal amplification: 25–37°C operation, no thermal cycler needed.
  • Ready-to-use premix for simplified reaction setup.
  • Enhanced specificity: lower background, more specific target DNA amplification.
  • Rapid amplification: DNA amplifiable in 5–10 min.
  • Cost-effective and accessible for diagnostics and research.
  • Flexible formats: liquid and lyophilized options available.
  • Diverse applications in healthcare, food safety, environmental monitoring.