Learn More
INTACT GENOMICS INC ig SYBR Green qPCR 2X Master Mix, 5000 Reactions, 10x5ml
Catalog No. NC3998169
Encompass
The SYBR Green qPCR 2X master mix is a ready-to-use cocktail for the amplification and detection of DNA in qPCR, containing all necessary components except primers and template.
  • Enhanced efficiency, specificity, and sensitivity
  • Compatibility with all major real-time PCR instruments
  • Superior gene expression results under various cycling conditions
  • Ready-to-use convenience
  • More affordable price
  • Includes an integrated chemically modified hot start Taq DNA polymerase, SYBR® Green I fluorescent dye, ROX dye, MgCl2, dNTPs, and stabilizers
  • Suitable for high-throughput real-time PCR screening and validation
  • High-quality hot start Taq DNA polymerase ensures higher fidelity and improved amplification
Catalog No. NC3998169 Supplier INTACT GENOMICS INC Supplier No. 3362
The SYBR Green qPCR 2X master mix is a ready-to-use cocktail for the amplification and detection of DNA in qPCR, containing all necessary components except primers and template.
  • Enhanced efficiency, specificity, and sensitivity
  • Compatibility with all major real-time PCR instruments
  • Superior gene expression results under various cycling conditions
  • Ready-to-use convenience
  • More affordable price
  • Includes an integrated chemically modified hot start Taq DNA polymerase, SYBR® Green I fluorescent dye, ROX dye, MgCl2, dNTPs, and stabilizers
  • Suitable for high-throughput real-time PCR screening and validation
  • High-quality hot start Taq DNA polymerase ensures higher fidelity and improved amplification